MultiLab Line BOD/pH Laboratory Kits
Kit for BOD/pH calculation
Manufacturer:  Ysi Incorporated YSI626574
 View more versions of this product
Catalog No. 09-324-569
Specifications Description & Specifications
Specifications
2
4010-2 Two channel benchtop instrument, optical BOD probe, double junction pH electrode and BOD analyst pro desktop software for calculation of BOD