YSI™ MultiLab Line BOD/pH Laboratory Kits
Kit for BOD/pH calculation
$4,860.47 - $5,987.40
Specifications
Channels 2
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
09-324-568 YSI™
YSI626573
4010-2 Two channel benchtop instrument, optical BOD probe and double junction pH electrode Each for $4,860.47
09-324-569 Ysi Incorporated
YSI626574
4010-2 Two channel benchtop instrument, optical BOD probe, double junction pH electrode and BOD analyst pro desktop software for calculation of BOD Each for $5,987.40
Specifications
Description
BOD/pH Lab Kit Includes
  • 4010-2 Two Channel benchtop instrument
  • Double Junction pH electrode
  • Optical BOD probe
Advanced BOD/pH Lab Kit Includes
  • 4010-2 Two Channel benchtop instrument
  • Double Junction pH electrode
  • Optical BOD probe
  • BOD Analyst Pro desktop software
Description & Specifications
Specifications
2