missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ Model 550A Dissolved Oxygen Meter

Rugged DO Meter designed for the field

Manufacturer:  YSI™ YSI550A50

 View more versions of this product

Catalog No. 13-298-122


 

Specifications Description & Specifications

Specifications

3 Years
Thermistor
±0.3°C
C Alkaline
CE, IP67
0.91kg
9 x 4.75 in. (23 x 12cm)
2.5 in.
50 ft.
0 to 50mg/L
0 to 500% Air Saturation
23° to 113°F
12cm
Meter, DO/temperature probe, 4 C Batteries, Cable
0.1°C
2000 hr.
67
2 lb.
0 to 20mg/L, ±0.3mg/L or 2% of reading, whichever is greater, 20 to 50mg/L, ±6% of reading
DO, Temperature
9 in.
23cm
0.01, 0.1mg/L
0.1% Air Saturation
4.75 in.