missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ Conductivity Calibration Standards

NIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy

Manufacturer:  YSI™ YSI065270

 View more versions of this product

Catalog No. 15-176-219



Description

Description

  • Conductivity solutions are inspected with reference to primary standard solutions according to OIML recommendation 56
  • Bottles include a table of corrections at temperatures between 20 and 30°C
  • Available as: 1 Quart or in a box of 8 individual pints

Specifications

Specifications

6.5 to 7.5
1g/mL
Traceable
Undesignated
1,000μS/cm (Fresh water)
1 quart
Liquid
100°C
0.9463L
Conduct calibration

Chemical Identifiers

0.0
icon-pdf.png
Documents