missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
YSI™ Colorimeter Reagents - Sulfate
$76.48 - $78.65
Specifications
Certifications/Compliance | EPA |
---|---|
For Use With (Application) | Used to measure sulfate |
For Use With (Equipment) | Photoflex colorimeter |
Catalog Number | Mfr. No. | Range | Volume (English) | Volume (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Range | Volume (English) | Volume (Metric) | Price | Quantity | |||
15-177-761
|
YSI™
251423Y |
2 to 70mg/L | 0.84 oz. | 25 mL |
Pack of 100 Each for $78.65
|
|
|||
15-177-760
|
YSI™
251413Y |
0 to 70mg/L | 0.33 oz. | 10 mL |
Pack of 100 Each for $76.48
|
|
|||
Specifications
EPA | |
Used to measure sulfate | |
Photoflex colorimeter |