YSI™ Colorimeter Reagents - Oxygen
Manufacturer:  YSI™ YSI251421Y
 View more versions of this product
Catalog No. 15-177-752
Specifications
Specifications
100/Pk.
0.004 to 0.45mg/L
Used to measure DEHA oxygen scavenger
DEHA1 TP
Powder
Photoflex colorimeter
Documents