YSI™ Colorimeter Reagents - Nitrogen
Manufacturer:  YSI™ YSI251995Y
 View more versions of this product
Catalog No. 15-177-803
Specifications
Specifications
50/Pk.
0.5 to 25mg/L
Used to measure total nitrogen
25.0
Nitrogen Ntot1 TC (Low range)
Vial
Photoflex colorimeter
Documents