YSI™ Colorimeter Reagents - Nitrite
Manufacturer:  YSI™ YSI251409Y
 View more versions of this product
Catalog No. 15-177-749
Specifications
Specifications
100/Pk.
0.002 to 0.3, 0.007 to 0.985mg/L
Used to measure nitrite
0.33 oz.
Nitrite NO2-1 TP (Low Range)
Powder
Photoflex colorimeter
10mL
Documents