missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ Colorimeter Reagents - Molybdate

$54.17 - $187.64

Specifications

Format Powder
For Use With (Equipment) Photoflex colorimeter
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Volume (English) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Volume (English) Volume (Metric) Price Quantity  
15-177-746 YSI™
YSI251407Y
Used to measure molybdate 0.33 oz. 10mL Pack of 100 Each for $154.75
15-177-747 YSI™
YSI251418Y
Used to measure molybdate 0.84 oz. 25mL Pack of 100 Each for $187.64
15-177-772 YSI™
YSI251407RP1Y
Used to measure molybdate, HR 1 replacement pack for molybdate Mo-1 TP 0.33 oz. 10mL Pack of 100 Each for $54.17
15-177-774 YSI™
YSI251407RP2Y
Used to measure molybdate, HR 2 replacement pack for molybdate Mo-1 TP 0.33 oz. 10mL Pack of 100 Each for $56.09
15-177-773 YSI™
YSI251418RP1Y
Used to measure molybdate, HR 1 replacement pack for molybdate Mo-2 TP 0.84 oz. 25mL Pack of 100 Each for $67.72
15-177-775 YSI™
YSI251407RP3Y
Used to measure molybdate, HR 3 replacement pack for molybdate Mo-1 TP 0.33 oz. 10mL Pack of 100 Each for $56.09
15-177-776 YSI™
YSI251418RP3Y
Used to measure molybdate, HR 3 replacement pack for molybdate Mo-2 TP 0.84 oz. 25mL Pack of 100 Each for $67.72
Specifications Description & Specifications

Specifications

Powder
Photoflex colorimeter