YSI™ Colorimeter Reagents - Manganese
Manufacturer:  YSI™ YSI251417Y
 View more versions of this product
Catalog No. 15-177-745
Specifications
Specifications
50/Pk.
0.007 to 0.7mg/L
Used to measure low range manganese
Manganese Mn-2 TP (Low Range)
Powder
Photoflex colorimeter
Documents