missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ Colorimeter Reagents - Iron

$36.72 - $315.04

Specifications

Format Powder
For Use With (Application) Used to measure total iron
For Use With (Equipment) Photoflex colorimeter
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Range Price Quantity  
Catalog Number Mfr. No. Range Price Quantity  
15-177-741 YSI™
YSI251404Y
0.012 to 1.8mg/L Pack of 100 Each for $54.11
15-177-742 YSI™
YSI251405Y
0.02 to 3mg/L Pack of 100 Each for $36.72
15-177-743 YSI™
YSI251405LPY
0.02 to 3mg/L Pack of 1000 Each for $315.04
Specifications Description & Specifications

Specifications

Powder
Used to measure total iron
Photoflex colorimeter