YSI™ Colorimeter Reagents - COD
Manufacturer:  YSI™ YSI251991LPY
 View more versions of this product
Catalog No. 15-177-790
Specifications
Specifications
150/Pk
20 to 1500mg/L
Photoflex colorimeter
COD2 TC (Mid Range)
Used to measure COD
Documents