YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
Supplier:  YSI™ 251397LPY
 View more versions of this product
Catalog No. 15-177-740
Description
Description
  • DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
Specifications
DPD
Powder
Used with other manufacturers colorimeter
0.169 oz.
5 mL
Used to measure total chlorine
0 to 3.5mg/L