YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
Supplier: YSI™ 251601Y
Description
- DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
EPA | |
Powder | |
0.01 to 6mg/L |
DPD No. 3 LR | |
Used to measure free chlorine |