YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
Manufacturer:  YSI™ YSI251603Y
 View more versions of this product
Catalog No. 15-177-810
Description
Description
  • DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
Specifications
EPA
0.1 to 10mg/L
Powder
Used to measure free chlorine
Documents