YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
Manufacturer:  YSI™ YSI251398Y
 View more versions of this product
Catalog No. 15-177-735
Description
Description
  • DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
Specifications
0 to 3.5mg/L
Used to measure free chlorine
0.169 oz.
Powder
Used with other manufacturers colorimeter
5mL
Documents