YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
Manufacturer:  YSI™ YSI251402LP1Y
 View more versions of this product
Catalog No. 15-177-734
Description
Description
  • DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
Specifications
0.05 to 5mg/L
Used to measure free chlorine
0.84 oz.
Powder
Photoflex or 900 colorimeter
25mL
Documents