missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
Supplier: YSI™ 251415Y
Description
- DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
DPD | |
Powder | |
Photoflex or 900 colorimeter | |
0.84 oz. |
25 mL | |
Used to measure total chlorine | |
0.05 to 5mg/L |