Specifications
EPA | |
Powder | |
Photoflex colorimeter | |
50/Pk. | |
Ammonium NH4-3 TC (HR) |
Nitrogen, Ammonia High Range, vial | |
Used to measure ammonium, HR | |
50 | |
0.4 to 50.0 mg/L NH4-N; 0.5 to 64.4 mg/L NH4; NH3-N |