Please login to your online account to display your discounted pricing
YSI™ Cables and DO/Temp/Conductivity Probes for 556 Multi-Probe System
Manufacturer: ysi™  YSI556320
 View more versions of this product
Catalog No. 13-298-72
  • / Each
Description & Specifications
Specifications
Warranty 1 Year
Length (English) Cable 64 ft.
Length (Metric) Cable 20m
Probe Type DO/temperature/conductivity probe
Includes Temperature/conductivity sensor
For Use With YSI model 556 handheld multi probe meter
Applications Measures DO, temperature, conductivity