missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ Buffer Solutions for pH Electrodes

Designed and manufactured to the highest quality standards

Manufacturer:  YSI™ YSI003821

 View more versions of this product

Catalog No. 15-176-213



Description

Description

  • Calibration buffer
  • For use with YSI 60, YSI 556, Ecosense™ pH10, Ecosense pH100, 6-Series sonde meters
  • Color coded: pH 4 (red), pH 7 (yellow), pH 10 (blue)

Specifications

Specifications

YSI™ 60, YSI™ 63, YSI™ 556, Ecosense™ pH10, Ecosense pH100, 6-series sonde meter
Red
4
Calibration Buffer
Documents