YSI™ 605790 Pro Plus Quatro Cable
Measure conductivity/temperature, dissolved oxygen and any two ISEs among pH, ORP, ammonium, nitrate or chloride
Manufacturer:  YSI™ YSI60579020
 View more versions of this product
Catalog No. 14-660-218
Description
Description
  • A true field cable with MS (military spec) connectorsR
  • ugged, weighted probe guard
Specifications
Specifications
Military Spec, Waterproof, Bayonet Lock
0.046 m
787.4 in.
1.85 in.
4-ports and 5560 temperature/conductivity sensor
20 m