YSI™ 6052030 Pro Series DO/Conductivity Cables
$1,128.32 - $1,519.60
Specifications
For Use With (Equipment) Professional Plus and Pro2030 DO/conductivity meter
Includes Built-in conductivity sensor and temperature/DO sensor port
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
15-177-490 YSI™
YSI60520304
157.4 in. 4m Each for $1,146.53
15-177-652 YSI™
YSI605203020
787.4 in. 20m Each for $1,387.09
13-398-94 YSI™
YSI60520301
39.3 in. 1m Each for $1,128.32
15-177-493 YSI™
YSI605203030
1181.1 in. 30m Each for $1,519.60
Specifications
Description
The 6052030 cables ONLY work with the YSI Pro Plus handheld
  • Measure dissolved oxygen, conductivity, salinity, specific conductance, total dissolved solids and temperature
  • Conductivity sensor is built into the cable
  • Automatic salinity compensation for DO readings
  • Cables do NOT come with the DO sensor
Description & Specifications
Specifications
Professional Plus and Pro2030 DO/conductivity meter
Built-in conductivity sensor and temperature/DO sensor port