Please login to your online account to display your discounted pricing
YSI™ 6052030 Pro Series DO/Conductivity Cables
Manufacturer: ysi™  YSI60520301
 View more versions of this product
Catalog No. 13-398-94
  • / Each
Description & Specifications
Specifications
Includes Built-in conductivity sensor and temperature/DO sensor port
Length (English) 39.3 in.
Length (Metric) 1m
For Use With (Equipment) Professional Plus and Pro2030 DO/conductivity meter