YSI™ 6052030 Pro Series DO/Conductivity Cables
Cables available in 4m, 10, 20, 30m lengths
Supplier:  YSI™ 605203010
 View more versions of this product
Catalog No. 15-177-491
Description
Description
The 6052030 cables ONLY work with the YSI Pro Plus handheld
  • Measure dissolved oxygen, conductivity, salinity, specific conductance, total dissolved solids and temperature
  • Conductivity sensor is built into the cable
  • Automatic salinity compensation for DO readings
  • Cables do NOT come with the DO sensor
Specifications
Specifications
Built-in conductivity sensor and temperature/DO sensor port
393.7 in.
10 m