YSI™ 6051030 Pro Series ISE/Conductivity Cable
Manufacturer:  YSI™ YSI605103010
 View more versions of this product
Catalog No. 15-177-487
Description
Description
Measures any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), conductivity, salinity, specific conductance, total dissolved solids and temperature
  • Conductivity sensor is built into the cable
  • Cables over 1-meter also come with the 603062 cable management kit
  • The 6051030 cables only work with the YSI Pro Plus handheld
  • Does not include ISE sensor 15-176-209 (order separately)
Specifications
Specifications
Conductivity, pH, Temp
393.7 in.
Conductivity sensor
Conductivity, pH, temperature meter
10m
Documents