YSI™ 6051030 Pro Series ISE/Conductivity Cable
$924.21 - $1,010.85
Specifications
For Use With (Equipment) Conductivity, pH, temperature meter
Includes Conductivity sensor
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
15-177-486 YSI™
YSI60510304
157.4 in. 4m Each for $924.21
15-177-487 YSI™
YSI605103010
393.7 in. 10m Each for $1,010.85
Specifications
Description
Measures any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), conductivity, salinity, specific conductance, total dissolved solids and temperature
  • Conductivity sensor is built into the cable
  • Cables over 1-meter also come with the 603062 cable management kit
  • The 6051030 cables only work with the YSI Pro Plus handheld
  • Does not include ISE sensor 15-176-209 (order separately)
Description & Specifications
Specifications
Conductivity, pH, temperature meter
Conductivity sensor