YSI™ 6051020 Pro Series ISE/DO Cable
$1,019.13 - $1,371.09
Specifications
For Use With (Equipment) Pro Plus DO/pH/temperature meter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
15-177-482 YSI™
YSI60510204
157.4 in. 4m Each for $1,168.47
15-177-483 YSI™
YSI605102010
393.7 in. 10m Each for $1,371.09
13-398-92 YSI™
YSI60510201
39.3 in. 1m Each for $1,019.13
Specifications
Description
  • Measure any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), dissolved oxygen and temperature
  • Cables over 1-meter also come with the 603062 cable management kit
  • The 6051020 cables ONLY work with the YSI Pro Plus handheld
  • DOES NOT come with the ISE sensor
Order Info
Order probes separately.
Description & Specifications
Specifications
Pro Plus DO/pH/temperature meter