YSI™ 60510 Pro Series ISE Cable
The 60510 is a 1-port cable for use with the Professional Plus instrument. The port requires a ISE probe.
Manufacturer:  YSI™ YSI6051030
 View more versions of this product
Catalog No. 15-177-469
Description
Description
Conductivity Probes and Multiparameter Cables. Order other probes separately.
  • The 60510 ISE (pH, ORP, pH/ORP, ammonium, nitrate, chloride) cables ONLY work with the YSI Pro Plus handheld
  • Any of the ISE sensors can be connected to this cable at any time
Specifications
Specifications
ProPlus handheld instrument
1181.1 in.
30m
Cable management kit
Documents