Please login to your online account to display your discounted pricing

YS-035 hydrochloride, Tocris


Inhibits K+ outward/pacemaker current

Quantity 10mg

Manufacturer: r & d systems  RDS041610

 View more versions of this product

Catalog No. 041610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 395.93
Assay Percent Range >98%
Chemical Name or Material YS-035 hydrochloride
Molecular Formula C21H29NO4·HCl
Recommended Storage Desiccate at Room Temperature
CAS 89805-39-0
Synonym N, N-Bis-(3, 4-dimethoxyphenylethyl)-N-methylamine hydrochloride

Specifications

Chemical Name or Material YS-035 hydrochloride
Assay Percent Range >98%
Molecular Formula C21H29NO4·HCl
Formula Weight 395.93
Synonym N, N-Bis-(3, 4-dimethoxyphenylethyl)-N-methylamine hydrochloride
Accessories

Desiccate at Room Temperature