missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ YPD Broth (Powder/Molecular Genetics)

$80.56 - $3,408.51

Specifications

Format Powder
Media Type YPD Broth
Grade Molecular genetics
Content And Storage 4°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BP2469-500
View Documents Promotion Details
Fisher BioReagents™
BP2469500
500 g
Each for $216.93
Case of 6 Each for $1,301.58
 
BP2469-100
View Documents Promotion Details
Fisher BioReagents™
BP2469100
100 g
Each for $80.56
Case of 6 Each for $483.36
 
BP2469-10
View Documents Promotion Details
Fisher BioReagents™
BP246910
10,000 g
Each for $3,408.51
 
Description

Description

Specifications

Specifications

Powder
YPD Broth
Molecular genetics
4°C
SDS
Product Certifications