Please login to your online account to display your discounted pricing

Youngstown Barrel & Drum Ultra-Utility Trays, Flexible Model™

Convenient, compact and foldable trays capture small leaks and spills

$83.76 - $83.76

Specifications

Color Yellow
Material PVC Fabric
Product Type Ultra Utility Tray™
For Use With (Application) Used to capture small leaks and spills
Includes Utility tie
View More Specs
Products
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Weight (English) Weight (Metric) Price Quantity    

19-171-367

 
youngstown barrel & drum
1335UT
1.5 gal., 5.7L 12 x 24 x 4.75 in. (30.48 x 60.96 x 12.06cm) 1 lb. 0.45kg Each for $83.76
Description & Specifications

Specifications

Color Yellow
Material PVC Fabric
Product Type Ultra Utility Tray™
For Use With (Application) Used to capture small leaks and spills
Includes Utility tie
Model Flexible

  • One piece units — no components or pieces to lose
  • High-visibility, 18 oz. PVC fabric provides rugged durability and excellent chemical compatibility
  • Simply unroll, then unfold — no straps, supports or sidewalls needed
  • Folds/rolls into small size
  • Utility tie provides easy and convenient storage and portability