Youngstown Barrel & Drum Ultra-Spill Pallets™
Built for heavy loads and convenient handling
$346.30 - $652.74
Specifications
Capacity 66 gal., 249.8L
Capacity (English) 66 gal.
Capacity (Metric) 249.8L
Certifications/Compliance SPCC, EPA
Material Polyethylene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Load Bearing Capacity (English) Model Product Type Price Quantity  
Catalog Number Mfr. No. Color Load Bearing Capacity (English) Model Product Type Price Quantity  
19-130-4777 Youngstown Barrel & Drum
1000UT
Yellow 6000 lb. P4 P4 Spill Pallet Each for $620.36
19-130-4778 Youngstown Barrel & Drum
1001UT
Yellow 6000 lb. P4 P4 Spill Pallet Each for $652.74
19-130-5023 Youngstown Barrel & Drum
2504UT
Black 1500 lb. P2 Economy P2 Economy Spill Pallet Each for $346.30
19-130-5025 Youngstown Barrel & Drum
1010UT
Yellow 3000 lb. P2 P2 Spill Pallet Each for $463.40
19-130-5026 Youngstown Barrel & Drum
1011UT
Yellow 3000 lb. P2 P2 Spill Pallet Each for $498.27
19-130-5024 Youngstown Barrel & Drum
2505UT
Black 1500 lb. P2 Economy P2 Economy Spill Pallet Each for $381.18
Specifications
Description
  • Combine high-strength and low-profile features
  • Highest load capacity Available 6000 lb. for four-Drum model, 3000 lb. for two-Drum model
  • Safety yellow sidewalls are translucent, offering convenient visual leak detection
  • Polyethylene construction compatible with broad range of containment for chemicals, including acids and corrosives
Compliance
Meet EPA and uniform fire code containment pallet regulations with Ultratech International pallets.
Description & Specifications
Specifications
66 gal., 249.8L
249.8L
Polyethylene
66 gal.
SPCC, EPA