Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Ultra-Sidewinder™ Cable Protection System: 3 Ft.
Protects cords and cables and avoid dangerous tripping hazards
$142.34 - $190.67
Specifications
Style 3-Foot System
View More Specs
Products
Catalog Number Mfr. No. Color Size Price Quantity    
18-938-520
youngstown barrel & drum
1830UT
Black and yellow Medium Each for $142.34
18-938-522
youngstown barrel & drum
1832UT
Black Medium Each for $142.34
18-938-524
youngstown barrel & drum
1840UT
Black and yellow Large Each for $190.67
18-938-526
youngstown barrel & drum
1842UT
Black Large Each for $190.67
Description & Specifications
Specifications
Style 3-Foot System
An innovative new solution for protecting cables run across a floor. The new design articulates left and right, allowing the system to turn corners and adapt to the layout of the room, while maintaining full protection for the cables and trip protection for pedestrians. Cables can be left in the system for the ultimate in portability, or left in place indefinitely.
  • Articulating pieces offer flexibility to design curves, corners and s-curves while maintaining completely flat profile
  • Modular design enables reconfigure, repair, and replace targeted segments, even while the cable remains connected.
  • Simple “snap in” assembly without cutting or using tools
  • Fully customizable length
  • Black and yellow color provides additional “safety tape” type visual warning
  • Also available in all black
  • Load Capacity: 260 lbs./ in.2 (18.3 kg./cm2)
  • Load Capacity per axle: 13,500 lbs. (6,123 kg)
  • Available in medium and large sizes
  • One-foot extensions available separately