missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Ultra-Overpack Plus™ 95 Poly Drum
Handles damaged or leaking containers of hazardous materials
$328.99 - $332.45
Specifications
Certifications/Compliance | UN 1H2\X345\S\09\USA\M5904 |
---|---|
Closure Type | Screw lid |
Color | Yellow |
Material | Plastic |
For Use With (Application) | For handles damaged or leaking containers of hazardous materials |
Catalog Number | Mfr. No. | Diameter (English) Outer | Height (English) | Weight (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Outer | Height (English) | Weight (English) | Price | Quantity | |||
19-171-522
|
Youngstown Barrel & Drum
0580UT |
25.5 to 32.25 in. | 41.25 in. | 48 lb. |
Each for $328.99
|
|
19-824-100
|
Youngstown Barrel & Drum
0582UT |
21 to 29 in. | 41.25 in. | 88.18 lb. |
Each for $332.45
|
|
Description
- Open head style
- Easy-to-close screw top lid
- Color: Yellow
- UN marking: 1H2\X345\S\09\USA\M5904
Order Info
Capacity: 95 gal. (360L). O.D. x H: 32.3 x 43.3 in. (82 x 110cm). Weight: 48 lb. (22kg).
Certifications
UN Transport of Dangerous Goods. IMO Dangerous Goods Code (IMDG) Annex I-Section 8. ICAO Technical Instructions for the Safe Transport of Dangerous Goods by Air, Part 7, Chapter 4. IATA Dangerous Goods Regulations, Section 10.3.
Compliance
Complies with UN packaging group 1 (X-Rated) and certified for use as DOT Salvage Drums, 49 CFR 173.3.
Specifications
UN 1H2\X345\S\09\USA\M5904 | |
Yellow | |
For handles damaged or leaking containers of hazardous materials | |
Open |
Screw lid | |
Plastic | |
50 |