Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Ultra-Hose Stand™
Keeps hoses off of berm sidewalls - helps maintain structural integrity and capacity of Containment Berm
$329.48 - $329.48
Specifications
Product Type Ultra Hose Stand
View More Specs
Products
Catalog Number Mfr. No. Model Material Price Quantity    
19-339-056
youngstown barrel & drum
8369UT
Heavy-Duty Polyurea Each for $329.48
Description & Specifications
Specifications
Product Type Ultra Hose Stand
  • Versatile unit that can be used completely inside or outside of Berm or straddling Berm with sidewall running through 8 in. wide opening in support sections
  • Heavy-duty, ribbed polyethylene support sections are compatible with fuel and many other chemicals
  • Unit quickly and easily disassembles into four flat pieces for easy transport and storage
  • Cutout in top of unit can handle up to 6.5 in. hoses and keeps them secure
  • Wide 27 in. dia. base keeps unit in place and upright
  • Dimensions: 27 × 27 × 23.875 in. (68.6 × 65.6 × 60.64cm)