Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Ultra-Hard Top Plus™ Models
Provide a safe, convenient and effective place to store drums
$6,054.16 - $9,006.49
Specifications
Capacity 55 gal., 208L
Certifications/Compliance SPCC, EPA, 40 CFR 264.175, NPDES, 40 CFR 122.26
Color Yellow
Material Polyethylene
Resistant Chemical
View More Specs
Products
Catalog Number Mfr. No. Holds Includes Weight (English) Product Type Price Quantity    
19-339-033
youngstown barrel & drum
9652UT
12 Drums No Drain 1190 lb. Ultra-Hard Top™ Plus P12 Spill Pallet Each for $8,976.58
19-339-032
youngstown barrel & drum
9651UT
8 Drums Drain 815 lb. Ultra-Hard Top™ Plus P8 Spill Pallet Each for $6,103.98
19-339-031
youngstown barrel & drum
9650UT
8 Drums No Drain 815 lb. Ultra-Hard Top™ Plus P8 Spill Pallet Each for $6,054.16
19-339-034
youngstown barrel & drum
9653UT
12 Drums Drain 1190 lb. Ultra-Hard Top™ Plus P12 Spill Pallet Each for $9,006.49
19-339-035
youngstown barrel & drum
9654UT
16 Drums No Drain 1565 lb. Ultra-Hard Top™ Plus P16 Spill Pallet CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-339-036
youngstown barrel & drum
9655UT
16 Drums Drain 1565 lb. Ultra-Hard Top™ Plus P16 Spill Pallet CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-339-038
youngstown barrel & drum
9657UT
20 Drums Drain 1940 lb. Ultra-Hard Top™ Plus P20 Spill Pallet CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-339-037
youngstown barrel & drum
9656UT
20 Drums No Drain 1940 lb. Ultra-Hard Top™ Plus P20 Spill Pallet CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Capacity 55 gal., 208L
Certifications/Compliance SPCC, EPA, 40 CFR 264.175, NPDES, 40 CFR 122.26
Color Yellow
Material Polyethylene
Resistant Chemical
All-polyethylene construction offers excellent chemical resistance and will not rust or corrode.
  • Low-profile containment pallet positions drum-tops at a safe, convenient level to pour wastes into funnels or to use drum pumps
  • Multiple 4-drum compartments offer flexible usage for a broad range of operations including: storage of virgin chemicals, active hazardous waste collection and storage of spill response supplies
  • Internal walls and separate sumps allow materials to be segregated
  • Swing-out doors provide safety and convenience for loading / unloading drums
  • Roll-top covers can be easily lifted from waist height to access drum tops
Meets EPA Container Storage Regulation 40 CFR 264.175 and helps comply with NPDES, 40 CFR 122.26 (1999). Also meets SPCC Regulations for Secondary Containment of hydrocarbons.