Please login to your online account to display your discounted pricing

Youngstown Barrel & Drum Ultra-Hard Top P1 Plus™

Will not rust or corrode

$762.37 - $762.37

Specifications

Material Polyethylene
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

19-339-029

 
youngstown barrel & drum
9640UT
Ultra-Hard Top™ Plus P1 Spill Pallet Unit Each for $762.37
Description & Specifications

Specifications

Material Polyethylene

“Roll-top“ cover allows quick access to drum pumps and funnels.

  • Tall head space (25.75 in. [65.4cm]) allows use with drum pumps and funnels
  • Constructed of 100% polyethylene
  • Can be moved by forklift under the “shoulder“ area or wheeled with dolly (not included)
  • Dimensions: 36 × 36 × 66 in. ( 91.4 x 91.4 x 167cm)

Unit load capacity: 800 lb. (363kg). Containment capacity: 64 gal. (242.3L). Weight: top, 36 lb. (16.3kg); bottom, 26 lb. (12kg).

Meets EPA Containment Storage Regulation 40CFR264.175. Helps meet Stormwater Management Regulations NPDES 40CFR122.26 (1999).