Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Ultra-Drum Tourniquets™
Latches onto drums to seal off leaks
$402.39 - $402.39
Specifications
For Use With (Application) Seal off puncture and corrosion leaks on drums and works on all surfaces including Polyethylene, Steel and Stainless Steel
View More Specs
Includes:  UV-resistant bag included
Products
Catalog Number Mfr. No. Weight (English) Weight (Metric) Price Quantity    
19-339-027
youngstown barrel & drum
2010UT
11 lb. 5kg Each for $402.39
Description & Specifications
Specifications
For Use With (Application) Seal off puncture and corrosion leaks on drums and works on all surfaces including Polyethylene, Steel and Stainless Steel
A chemical-resistant sealing pad is compressed against the damaged area to stop leaks on contact.
  • 8.9×17.8cm (3.5×7 in.) maximum sealing area
  • 305cm (10 ft.) long ratchet strap
  • No tools required
  • Magnetic model has powerful magnets that latch onto steel drums to seal off puncture and corrosion leaks. A chemical-resistant sealing pad is compressed against the damaged area to stop leaks on contact
  • Universal model features a ratchet strap system. Works on all surfaces including polyethylene, steel and stainless steel
For temporary use only