Please login to your online account to display your discounted pricing

Youngstown Barrel & Drum Ultra-Drain Seal Plus™

Specially rugged reinforced PVC top layer provides protection from extended UV exposure and vehicular traffic

$204.72 - $1,147.36

Specifications

Certifications/Compliance NPDES, SPCC
Color Bright Yellow
Material Urethane, PVC
Product Type Ultra-Drain Seal™ Plus
For Use With (Application) Used in parking areas, loading docks and other areas where drains may be driven over
View More Specs
Products
Catalog Number Mfr. No. Shape Weight (English) Weight (Metric) Price Quantity    

19-171-054

 
youngstown barrel & drum
2160UT
Square 23 lb. Each for $643.53

19-171-053

 
youngstown barrel & drum
2167UT
Square 15 lb. 6.8kg Each for $304.29

19-171-058

 
youngstown barrel & drum
2174UT
Circular 4 lb. 2kg Each for $204.72

19-171-055

 
youngstown barrel & drum
2161UT
Square 27 lb. 12.2kg Each for $822.04

19-171-052

 
youngstown barrel & drum
2166UT
Square 7 lb. 3.17kg Each for $232.16

19-171-059

 
youngstown barrel & drum
2175UT
Circular 7 lb. 3.17kg Each for $283.17

19-171-056

 
youngstown barrel & drum
2162UT
Square 43 lb. 19.5kg Each for $984.72

19-171-060

 
youngstown barrel & drum
2176UT
Circular 13 lb. 6kg Each for $537.97

19-171-062

 
youngstown barrel & drum
2164UT
Rectangle 20 lb. 9kg Each for $633.73

19-171-061

 
youngstown barrel & drum
2177UT
Circular 26 lb. 12kg Each for $735.93

19-171-063

 
youngstown barrel & drum
2165UT
Rectangle 38 lb. 17kg Each for $1,147.36
Description & Specifications

Specifications

Certifications/Compliance NPDES, SPCC
Color Bright Yellow
Material Urethane, PVC
Product Type Ultra-Drain Seal™ Plus
For Use With (Application) Used in parking areas, loading docks and other areas where drains may be driven over
Resistant Oil, water and most aggressive chemicals

  • Urethane bottom layer (same as Ultra-Drain seal) keeps spills from entering drains
  • For use in parking areas, loading docks and other areas where drains may be driven over
  • Bright yellow color helps indentify protected drains