Please login to your online account to display your discounted pricing

Youngstown Barrel & Drum Ultra-Containment Berms™, Ultimate Models

With unique “living hinge” design

$9,579.41 - $9,625.87

Specifications

Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Color Black
Material Copolymer
Product Type Ultra-Containment Berm™
For Use With (Application) Allows walls to be lowered as needed for placing products into containment areas, while still allowing vehicles to drive directly over entrance and exit walls of the berm without any effort
View More Specs
Products
Catalog Number Mfr. No. Length (English) Weight (English) Width (English) Price Quantity    

19-339-012

 
youngstown barrel & drum
8505UT
60 in. 681 lb. 12 in. Each for $9,625.87

19-339-013

 
youngstown barrel & drum
8506UT
50 in. 719 lb. 15 in. Each for $9,579.41
Description & Specifications

Specifications

Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Color Black
Material Copolymer
Product Type Ultra-Containment Berm™
For Use With (Application) Allows walls to be lowered as needed for placing products into containment areas, while still allowing vehicles to drive directly over entrance and exit walls of the berm without any effort
For Use With (Equipment) Vehicles, tankers, drums, tanks, machinery, and several other facility or construction site equipment
Height (English) 1 ft.
Height (Metric) 30.48cm
Includes Collapsible PVC sidewall assemblies, swivel to lower the sidewalls
Model Ultimate

  • Standard material of construction is Copolymer-2000™
  • XR5, polyurethane and other materials also available
  • Drive in/Drive Out end walls, once a vehicle has entered or exited, walls automatically spring back to their vertical position
  • Entry and exit walls are supported every 12 in. with proprietary “living hinge”
  • Both long sidewalls are held upright by PVC sidewall assemblies
  • Swivel feet to lower the sidewalls for storage or transport to another
  • For drums, IBCs, vehicles, equipment, frac tanks and other large containers