Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Ultra-Containment Berms™, Rapid Rise Model
Sidewalls stay lowered and out of the way when not in use
$1,153.53 - $9,081.23
Specifications
Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Material Copolymer 2000™
Product Type Rapid Rise Ultra-Containment Berm™
For Use With (Application) Used for normal operations, loading and unloading
Height (English) 1 ft.
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Length (English) Weight (English) Width (English) Price Quantity    
19-171-089
youngstown barrel & drum
8431UT
269 gal. 6 ft. 35 lb. 6 ft. Each for $1,337.88
19-171-088
youngstown barrel & drum
8430UT
179 gal. 6 ft. 29 lb. 4 ft. Each for $1,153.53
19-171-090
youngstown barrel & drum
8432UT
748 gal. 10 ft. 63 lb. 10 ft. Each for $2,326.98
19-171-091
youngstown barrel & drum
8437UT
2333 gal. 26 ft. 149 lb. 12 ft. Each for $4,900.63
19-171-093
youngstown barrel & drum
8434UT
5610 gal. 66 ft. 357 lb. 15 ft. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-171-092
youngstown barrel & drum
8433UT
5610 gal. 50 ft. 278 lb. 15 ft. Each for $9,081.23
Description & Specifications
Specifications
Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Material Copolymer 2000™
Product Type Rapid Rise Ultra-Containment Berm™
For Use With (Application) Used for normal operations, loading and unloading
Height (English) 1 ft.
Height (Metric) 0.3m
Vehicles and equipment can drive in and out with no set-up or take-down required. In the case of a spill, the foam ring around the top perimeter of the Containment Berm rises with the level of spilled liquid.
  • Helps meet EPA Container Storage Regulation 40 CFR 264.175 and SPCC requirements
  • Copolymer 2000™ construction