missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Tight Head Steel Drum
Youngstown Tight Head Steel Pail is perfect for the transporting of hazardous solids or liquids.
Supplier: Youngstown Barrel & Drum 457L
Description
With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.
- Salvage drum designed to overpack and ship damaged and/or leaking drums
- Also qualify for direct containment of hazardous solids
Specifications
Lined Drum | |
Bung | |
30 gal. | |
Black | |
Carbon Steel | |
Tight | |
29 in. | |
18.25 in. | |
24 lb. |
UN 1A1/Y1.8/250-DOT/UN | |
20/18 Gauge | |
114 L | |
For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums | |
50 | |
Ea. | |
73.66 cm | |
46.35 mm | |
10.88 Kg |