missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Youngstown Barrel & Drum Plastic Open-Head Drums

High-density polyethylene containers for transport of hazardous solids or liquids

Manufacturer:  Youngstown Barrel & Drum 1746

 View more versions of this product

Catalog No. 19-120-004



Description

Description

  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking

Specifications

Specifications

UN 3H2/Y90/S
4.33cm
50
24 in.
Black lid
4.85 lb.
15 gal.
Black,Blue
11 in.
For transport of hazardous solids or liquids
Open
60.96cm
“Euro” Style Drum
2.19kg
56.78L
Plastic
Documents