Please login to your online account to display your discounted pricing
Youngstown Barrel and Drum Oil and Debris Blocker
Blocker prevents pollutants from leaving the storm drain
$126.81 - $302.41
Specifications
Certifications/Compliance NPDES, 40 CFR 122.26, TMDL
Material Polyethylene
Product Type Ultra Oil And Debris Blocker™
For Use With (Application) Stop pollutants before they can reach stormwater outflow
For Use With (Equipment) Installed in stormwater outflows
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Weight (English) Price Quantity    
19-171-115
youngstown barrel & drum
9380UT
12 x 4.5 x 14 in. (30.5 x 11.5 x 35.6cm) 4 lb. Each for $302.41
19-171-122
youngstown barrel & drum
9387UT
12 x 11 x 12 in. (30.5 x 27.94 x 30.48cm) 3 lb. Each for $146.33
19-171-121
youngstown barrel & drum
9386UT
10 x 9.5 x 12 in. (25.4 x 24.13 x 30.48cm) 2 lb. Each for $126.81
Description & Specifications
Specifications
Certifications/Compliance NPDES, 40 CFR 122.26, TMDL
Material Polyethylene
Product Type Ultra Oil And Debris Blocker™
For Use With (Application) Stop pollutants before they can reach stormwater outflow
For Use With (Equipment) Installed in stormwater outflows
  • Water is forced through the bottom of the unit, leaving pollution floating or sinking
  • Will not corrode
  • Easily installed
  • NPDES
  • 40 CFR 122.26 (1999)
  • TMDL