Please login to your online account to display your discounted pricing

Youngstown Barrel & Drum Modular IBC Spill Pallets™, Outdoor

$6,650.57 - $9,413.72

Specifications

Color Yellow
Material Polyethylene
For Use With (Application) Allow convenient loading and unloading of IBC tank
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Certifications/Compliance Dimensions (L x W x H) Product Type Price Quantity    

19-160-0128

 
youngstown barrel & drum
1165UT
280 gal. NPDES, 40 CFR 122.26 126 x 93 x 79 in. (320.04 x 236.22 x 200.6cm) 2-Tank Outdoor Each for $6,650.57

19-160-0130

 
youngstown barrel & drum
1167UT
365 gal. 40 CFR 264.175, SPCC, EPA 250 x 93 x 79 in. (635 x 236.22 x 200.66cm) 4-Tank Outdoor Ultra- Modular Ibc Spill Pallet CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

19-160-0129

 
youngstown barrel & drum
1166UT
355 gal. 40 CFR 264.175, SPCC, EPA 188 x 93 x 79 in. (477.52 x 236.22 x 200.66cm) 3-Tank Outdoor Ultra- Modular Ibc Spill Pallet Each for $9,413.72

19-160-0131

 
youngstown barrel & drum
1168UT
375 gal. 40 CFR 264.175, SPCC, EPA 312 x 62 x 79 in. (792.48 x 157.48 x 200.66cm) 5-Tank Outdoor Ultra- Modular Ibc Spill Pallet CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Yellow
Material Polyethylene
For Use With (Application) Allow convenient loading and unloading of IBC tank