missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Youngstown Barrel & Drum Ultra-Spill Pallets Plus™

Low-profile design—only 8.75 in. high (22.2cm)

Manufacturer:  Youngstown Barrel & Drum 9610UT

 View more versions of this product

Catalog No. 19-130-5027



Description

Description

  • Nearly 50% lower than standard spill pallets for safer and less strenuous handling
  • Removable grates feature non-slip surface
  • All-polyethylene construction
  • Forkliftable

Order Info

Available in one-, two-, three- and four-drum configurations, with or without a drain

Compliance

Meet U.S. EPA container storage regulation 40 CFR 264.175, Uniform Fire Code spill containment regulations for use with drums and is SPCC compliant.

Specifications

Specifications

66 gal.
66 gal., 249.8L
Yellow
Reduces the potential for hazardous fluids to splash into the eyes or face
22.22cm
65.5 in.
4500 lb.
P2
40kg
101.6cm
Without drain
249.8L
SPCC, EPA, 40 CFR 264.175, UFC
65.5 x 40 x 8.75 in. (166.3 x 101.6 x 22.22cm)
8.75 in.
2 Drums
166.3cm
2041kg
88 lb.
40 in.
Polyethylene
Documents