Youngstown Barrel & Drum Ultra-Burp-Free Funnel™
Built-in vent eliminates splashing and “burping” of liquids while pouring
Manufacturer:  Youngstown Barrel & Drum 0651
 View more versions of this product
Catalog No. 19-058-090
Description
Description
Rust-free, noncorroding polyethylene construction provides superior chemical resistance.
  • Hinged lid may be locked to prevent unauthorized access
  • With threaded brass insert screws to fit a 2 in. (5.1cm) drum bung
Specifications
Specifications
27.9cm
Built-in vent, hinged lid, threaded brass insert
PE (Polyethylene)