missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YM 976, Tocris

PDE4 inhibitor

$181.19 - $762.58

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C17H16ClN3O
Formula Weight 313.79
Synonym 4-(3-Chlorophenyl)-1, 7-diethylpyrido[2, 3-d]pyrimidin-2(1H)-one
Chemical Name or Material YM 976
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
182110 R & D Systems
182110
10mg Each for $181.19
182150 R & D Systems
182150
50mg Each for $762.58
Specifications Description & Specifications

Chemical Identifiers

>98%
313.79
YM 976
C17H16ClN3O
4-(3-Chlorophenyl)-1, 7-diethylpyrido[2, 3-d]pyrimidin-2(1H)-one

Specifications

313.79
191219-80-4
C17H16ClN3O
YM 976
10mg
4-(3-Chlorophenyl)-1, 7-diethylpyrido[2, 3-d]pyrimidin-2(1H)-one
>98%

Safety and Handling

Recommended Storage : Store at Room Temperature