Please login to your online account to display your discounted pricing

YM 750, Tocris

Acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material YM 750
Assay Percent Range >98%
Molecular Formula C31H36N2O
Formula Weight 452.63
Synonym N-Cycloheptyl-N-(9H-fluoren-2-ylmethyl)-N'-(2, 4, 6-trimethylphenyl)urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

303910

 
r & d systems
RDS303910
10mg Each for $188.60

303950

 
r & d systems
RDS303950
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 452.63
Quantity 10mg
Chemical Name or Material YM 750
Synonym N-Cycloheptyl-N-(9H-fluoren-2-ylmethyl)-N'-(2, 4, 6-trimethylphenyl)urea
Assay Percent Range >98%
Molecular Formula C31H36N2O
CAS 138046-43-2
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C